Skip to Content
Merck
All Photos(1)

Key Documents

12-1450

Sigma-Aldrich

Gold(III) chloride hydrate

SAJ special grade, ≥99.0%

Synonym(s):

Auric chloride, Chloroauric acid, Hydrogen tetrachloroaurate(III) hydrate, Tetrachloroauric(III) acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HAuCl4 · aq
CAS Number:
Molecular Weight:
339.79 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
Assay:
≥99.0%
grade:
SAJ special grade
form:
solid
Pricing and availability is not currently available.

grade

SAJ special grade

Assay

≥99.0%

form

solid

availability

available only in Japan

density

3.9 g/mL at 25 °C

SMILES string

O.Cl.Cl[Au](Cl)Cl

InChI

1S/Au.4ClH.H2O/h;4*1H;1H2/q+3;;;;;/p-3

InChI key

OVJUDSKPNIBLOO-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Application


  • Etlingera elatior-Mediated Synthesis of Gold Nanoparticles and Their Application as Electrochemical Current Enhancer.: This study explores the synthesis of gold nanoparticles using Etlingera elatior extract, demonstrating the potential for these nanoparticles, likely including Gold(III) chloride hydrate, in enhancing electrochemical currents, relevant for sensors and diagnostic devices (Azri et al., 2019).


Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B - STOT RE 2 Oral

Target Organs

Kidney

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service