Skip to Content
Merck
All Photos(1)

Key Documents

PF125

Sigma-Aldrich

BACE1, His•Tag®, Human, Recombinant, NSO Cells

Recombinant, human BACE-1 (amino acids 1-460) fused at the C-terminus to a His•Tag sequence and expressed in NSO cells. Supplied as a mixture of ~68-70 kDa proenzyme and ~65-67 kDa mature enzyme.

Synonym(s):

Human β-Secretase

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202

recombinant

expressed in NSO cells

Quality Level

Assay

≥90% (SDS-PAGE)

form

lyophilized

specific activity

≥1 pmol/min-μg protein

does not contain

preservative

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

impurities

≤1 EU/μg Endotoxin (EU/μg BACE-1)

solubility

water: 500 μg/mL

shipped in

wet ice

storage temp.

−20°C

General description

as measured by the cleavage of 10 μM MCA-Ser-Glu-Val-Asn-Leu-Asp-Ala-Glu-Phe-Arg-Lys(DNP)-Arg-Arg-NH2 fluorescent substrate using 100 ng BACE-1 in 0.1 M sodium acetate (pH 4.0) at room temperature

Warning

Toxicity: Standard Handling (A)

Physical form

Lyophilized from sterile-filtered PBS.

Reconstitution

Following reconstitution refrigerate (4°C) for short-term storage or aliquot and freeze (-20°C or -70°C) for long-term storage. Stock solutions are stable for up to 1 month at 4°C or for up to 4 months at -20°C or -70°C. Avoid freeze/thaw cycles of solutions.

Other Notes

Ermolieff, J. et al. 2000. Biochemistry39, 12450.
Lin, X. et al. 2000. Proc. Natl. Acad. Sci. USA97, 1456.
Sinha, S. et al. 1999. Nature402, 537.
Vassar, R. et al. 1999. Science286, 735.
Yan, R. et al. 1999. Nature402, 533.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
HIS TAG is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Amanda Gonzales et al.
Journal of neuroscience methods, 202(1), 70-76 (2011-09-06)
The aspartyl protease BACE1 is the rate limiting enzyme in the synthesis of amyloid beta, which accumulation in the human brain is a hallmark of Alzheimer's disease (AD). BACE1 has been proposed as a surrogate marker of AD; however, very

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service