PC16
Anti-FGF, Basic (Ab-2) (40-63) Rabbit pAb
liquid, Calbiochem®
Synonym(s):
Anti-Fibroblast Growth Factor, Basic, Anti-FGF-2, Anti-Heparin Binding Growth Factor, Anti-HBGF-1
About This Item
Recommended Products
biological source
rabbit
Quality Level
antibody form
purified antibody
antibody product type
primary antibodies
clone
polyclonal
form
liquid
does not contain
preservative
species reactivity
human, rat
manufacturer/tradename
Calbiochem®
storage condition
OK to freeze
avoid repeated freeze/thaw cycles
isotype
IgG
shipped in
wet ice
storage temp.
−20°C
target post-translational modification
unmodified
Gene Information
human ... FGF2(2247)
General description
Immunogen
Application
Paraffin Sections (1-5 µg/ml, no pre-treatment required)
Packaging
Warning
Physical form
Reconstitution
Analysis Note
Fetal liver or kidney tissue
Other Notes
Fiege, J.J. and Baird, A. 1989. Proc. Natl. Acad. Sci.86, 3174.
Florkiewicz, R.Z. and Sommer, A. 1989. Proc. Natl. Acad. Sci.86, 3978.
Rifkin, D.B. and Moscatelli, D. 1989. J. Cell Biol.109, 1.
Baird, A., et al. 1988. Proc. Natl. Acad. Sci.85, 2324.
Chodak, G.W., et al. 1988. Cancer Res. 48, 2083.
Sommer, A., et al. 1987. Res. Comm.144, 543.
Abraham, J.A., et al.1986. EMBO J.5, 2523.
Esch, F., et al.1985. Proc. Natl. Acad. Sci.82, 6507.
Knighton, D., et al. 1977. Br. J. Cancer35, 347.
Legal Information
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service