Skip to Content
Merck
All Photos(1)

Key Documents

8.14565

Sigma-Aldrich

di-Rhenium decacarbonyl

for synthesis

Synonym(s):

di-Rhenium decacarbonyl

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Re2(CO)10
CAS Number:
Molecular Weight:
652.52
MDL number:
UNSPSC Code:
12161603
EC Index Number:
238-202-8
Pricing and availability is not currently available.

form

crystals

mp

181-184 °C

storage temp.

2-30°C

InChI

1S/10CO.2Re/c10*1-2;;

InChI key

ZIZHEHXAMPQGEK-UHFFFAOYSA-N

Analysis Note

Assay (ex C): ≥ 99.0 %
Identity (IR): passes test

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation

Storage Class Code

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service