Documents
1.17318
Millipore
RapidMicroBio Perforated Lid
hole D 0.7 mm, no. of holes, 400 autoclavable, aluminum lid, for use with MAS-100 NT®, for use with MAS-100 VF®
About This Item
UNSPSC Code:
41103403
NACRES:
NA.74
material
aluminum lid
Agency
non-compendial
feature
no. of holes 400 autoclavable
hole D
0.7 mm
application(s)
air monitoring
cosmetics
food and beverages
pharmaceutical
compatibility
for use with Contact Plates
for use with MAS-100 NT®
for use with MAS-100 VF®
General description
The RapidMicroBio Perforated Lid with 400x0.7mm holes is designed for use with MAS-100® portable air samplers MAS-100 VF®, MAS-100 NT® /MAS-100 NT® Ex or MAS-100 Eco® systems. It is compatible with environmental monitoring cassettes of the Growth Direct® System. This perforated lid is made from anodized aluminum and can be autoclaved. It provides a nominal d50 of 1.6 μm at a flow rate of 100 litre per minute.
For all microbial air samplers except MAS-100 VF® device specific clamps are required to center the cassettes. They are already included in the scope of delivery of article 1.19166.
Application
The RapidMicroBio Perforated Lid 400x0.7mm holes, aluminum for MAS-100® microbial air samplers NT/NT Ex VF/Eco is used for microbial air monitoring using Growth Direct® environmental monitoring cassettes.
Legal Information
Growth Direct is a registered trademark of Rapid Micro Biosystems, Inc.
MAS-100 is a registered trademark of MBV AG
MAS-100 Eco is a registered trademark of MBV AG
MAS-100 NT is a registered trademark of MBV AG
MAS-100 VF is a registered trademark of MBV AG