Skip to Content
Merck
All Photos(1)

Key Documents

880341P

Avanti

16:0-16:0 (16-F) PC

Avanti Research - A Croda Brand 880341P, powder

Synonym(s):

1-palmitoyl-2-(16-fluoropalmitoyl)-sn-glycero-3-phosphocholine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C40H79NO8PF
CAS Number:
Molecular Weight:
752.03
UNSPSC Code:
12352211
NACRES:
NA.25
Pricing and availability is not currently available.

form

powder

packaging

pkg of 1 × 10 mg (880341P-10mg)
pkg of 1 × 25 mg (880341P-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 880341P

shipped in

dry ice

storage temp.

−20°C

SMILES string

[O-]P(OCC[N+](C)(C)C)(OC[C@]([H])(OC(CCCCCCCCCCCCCCC([H])([H])F)=O)COC(CCCCCCCCCCCCCCC)=O)=O

General description

16:0-16:0 (16-F) PC or 1-palmitoyl-2-(16-fluoropalmitoyl)-sn-glycero-3-phosphocholine is a monofluorinated analog of 1,2-dipalmitoyl-sn-glycero-3-phosphocholine (DPPC).[1]

Application

16:0-16:0 (16-F) PC or 1-palmitoyl-2-(16-fluoropalmitoyl)-sn-glycero-3-phosphocholine is suitable for use as a rotational-echo double-resonance (REDOR) probe to analyze peptides integrated into phospholipid bilayers.[1]

Biochem/physiol Actions

Fluorinated lipids act as drug carriers. They aid in characterizing proteins and membranes. 16:0-16:0 (16-F) PC or 1-palmitoyl-2-(16-fluoropalmitoyl)-sn-glycero-3-phosphocholine automatically interdigitates lipid bilayers without inducing agents.[2] This is due to its hydrophilic nature because of the presence of highly polar C-F bond at the terminal end of the sn-2 acyl chain.[1][2]

Packaging

5 mL Amber Glass Screw Cap Vial (880341P-10mg)
5 mL Amber Glass Screw Cap Vial (880341P-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eric A Smith et al.
Chemistry and physics of lipids, 165(2), 151-159 (2011-12-28)
Unlike the parent phospholipid, 1,2-dipalmitoyl-sn-glycero-3-phosphocholine (DPPC), the monofluorinated analog, 1-palmitoyl-2-(16-fluoropalmitoyl)sn-glycero-3-phosphocholine (F-DPPC), spontaneously forms an interdigitated gel phase (L(β)I) below the main transition temperature (T(m)). We have examined the effects of introducing cholesterol to F-DPPC and 1:1 F-DPPC/DPPC membranes using a
Eric A Smith et al.
Biophysical chemistry, 147(1-2), 20-27 (2010-01-13)
In aqueous solution, the monofluorinated phospholipid 1-palmitoyl-2-[16-fluoropalmitoyl]sn-glycero-3-phosphocholine (F-DPPC) interdigitates without the use of inducing agents. To understand the thermal and physical properties of this unique lipid, F-DPPC was combined with the non-fluorinated 1,2-dipalmitoyl-sn-glycero-3-phosphocholine (DPPC), and 1,2-diarachidoyl-sn-glycero-3-phosphocholine (DAPC). Differential scanning calorimetry
Syed W H Shah et al.
Chemistry and physics of lipids, 230, 104918-104918 (2020-05-18)
The monolayer behavior of a l-DPPC derivative with a single fluorination in one of its terminal methyl groups (F-DPPC) at air-water interface was investigated by epifluorescence microscopy and infrared reflection absorption spectroscopy (IRRAS). Epifluorescence microscopy was utilized to study the
Babak Sanii et al.
The journal of physical chemistry. B, 114(1), 215-219 (2009-12-17)
In free bilayers, the fluid to gel main phase transition of a monofluorinated phospholipid (F-DPPC) transforms a disordered fluid bilayer into a fully interdigitated monolayer consisting of ordered acyl tails. This transformation results in an increase in molecular area and
D J Hirsh et al.
Biophysical journal, 75(4), 1858-1868 (1998-09-24)
16-Fluoropalmitic acid was synthesized from 16-hydroxypalmitic acid using diethylaminosulfur trifluoride. This monofluorinated fatty acid then was used to make 1-palmitoyl-2-[16-fluoropalmitoyl]-phosphatidylcholine (F-DPPC) as a fluorinated analog of dipalmitoylphosphatidylcholine (DPPC). Surprisingly, we found that the phase transition temperature (Tm) of F-DPPC occurs

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service