Key Documents
TEM-CF200NI
Sigma-Aldrich
Carbon Film Supported Nickel Square Mesh
size 200 mesh, box of 25
Synonym(s):
CARBON FILM 200 MESH NI 25/BX
About This Item
UNSPSC Code:
41111761
Pricing and availability is not currently available.
packaging
box of 25
size
200 mesh
General description
A 5-6 nm thin film of pure carbon supported standard squre mesh grids. Nickel grid, 200 mesh.
Choose from one of the most recent versions:
Certificates of Analysis (COA)