Key Documents
TEM-CF150NI
Sigma-Aldrich
Carbon Film Supported Nickel Square Mesh
size 150 mesh, box of 25
Synonym(s):
CARBON FILM 150 MESH NI
About This Item
UNSPSC Code:
41111761
packaging
box of 25
size
150 mesh
General description
A 5-6 nm thin film of pure carbon supported standard squre mesh grids. Nickel grid, 150 mesh.
Choose from one of the most recent versions:
Certificates of Analysis (COA)