Skip to Content
Merck
All Photos(1)

Key Documents

P23989

Sigma-Aldrich

N,N′-(1,4-Phenylene)dimaleimide

97%

Synonym(s):

1,4-Dimaleimidobenzene, N,N′-(p-Phenylene)dimaleimide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H8N2O4
CAS Number:
Molecular Weight:
268.22
Beilstein:
249631
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
Pricing and availability is not currently available.

Assay

97%

form

powder

mp

>300 °C (lit.)

SMILES string

O=C1C=CC(=O)N1c2ccc(cc2)N3C(=O)C=CC3=O

InChI

1S/C14H8N2O4/c17-11-5-6-12(18)15(11)9-1-2-10(4-3-9)16-13(19)7-8-14(16)20/h1-8H

InChI key

AQGZJQNZNONGKY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M O Steinmetz et al.
The Journal of cell biology, 138(3), 559-574 (1997-08-11)
The effect of the type of metal ion (i.e., Ca2+, Mg2+, or none) bound to the high-affinity divalent cation binding site (HAS) of actin on filament assembly, structure, and dynamics was investigated in the absence and presence of the mushroom
Ryan N Mello et al.
Biophysical journal, 102(5), 1088-1096 (2012-03-13)
We have used thiol cross-linking and electron paramagnetic resonance (EPR) to resolve structural transitions of myosin's light chain domain (LCD) and catalytic domain (CD) that are associated with force generation. Spin labels were incorporated into the LCD of muscle fibers
K Polosukhina et al.
Biochemistry, 36(39), 11952-11958 (1997-10-08)
Rate constants for the reactions of Cys-697 and Cys-707 of skeletal muscle myosin subfragment 1 (S1) with N,N'-p-phenylenedimaleimide (pPDM) and its monofunctional analog phenylmaleimide (PM) were measured for S1 and S1 bound to nucleotides and/or actin. The [pPDM] and [PM]
Agnieszka Galińska-Rakoczy et al.
Journal of molecular biology, 387(4), 869-882 (2009-04-03)
The mechanism of salt-induced actin polymerization involves the energetically unfavorable nucleation step, followed by filament elongation by the addition of monomers. The use of a bifunctional cross-linker, N,N'-(1,4-phenylene)dimaleimide, revealed rapid formation of the so-called lower dimers (LD) in which actin
Yoshitaka Kimori et al.
The Biochemical journal, 450(1), 23-35 (2012-12-06)
In the present paper, we described our attempt to characterize the rough three-dimensional features of the structural analogue of the key intermediate of myosin's cross-bridge cycle. Using quick-freeze deep-etch replica electron microscopy, we observed that actin-attached myosin during in vitro

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service