Skip to Content
Merck
All Photos(2)

Documents

DWKWGW541VTMLA

Azlon Multi-Lingual Safety Wash Bottles With Driplok Vapor Venting

label, Ethanol (in English, French, Spanish), capacity 500 mL

Synonym(s):

WGW541VTMLA

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41103209
NACRES:
NB.43

feature

label Ethanol (in English, French, Spanish)

packaging

pkg of 5 ea

manufacturer/tradename

Azlon WGW541VTMLA

capacity

500 mL

Looking for similar products? Visit Product Comparison Guide

Application

Ideal for ethanol dispensing within the laboratory

Features and Benefits

Multilingual with solvent labels including all new globally harmonised symbols, NFPA diamond, risk phrases and safety information

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service