All Photos(2)
About This Item
Linear Formula:
FC6H4C6H4F
CAS Number:
Molecular Weight:
190.19
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
97%
form
crystals
bp
254-255 °C (lit.)
mp
88-91 °C (lit.)
SMILES string
Fc1ccc(cc1)-c2ccc(F)cc2
InChI
1S/C12H8F2/c13-11-5-1-9(2-6-11)10-3-7-12(14)8-4-10/h1-8H
InChI key
PZDAAZQDQJGXSW-UHFFFAOYSA-N
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
David Hughes et al.
Biodegradation, 22(4), 741-749 (2010-09-11)
Fluorinated aromatic compounds are significant environmental pollutants, and microorganisms play important roles in their biodegradation. The effect of fluorine substitution on the transformation of fluorobiphenyl in two bacteria was investigated. Pseudomonas pseudoalcaligenes KF707 and Burkholderia xenovorans LB400 used 2,3,4,5,6-pentafluorobiphenyl and
L van Bree et al.
Journal of biochemical toxicology, 5(1), 57-63 (1990-01-01)
The effects of pretreatment with symmetrically dihalogenated biphenyls (DXBs, X-F, Cl(C), Br(B) and I) on rat liver drug metabolism enzymes were investigated. 4,4'-DFB, -DCB, and -DBB as well as 2,2'-DFB appeared to be inducers of microsomal cytochrome P-450-linked monoxygenases (N-demethylases
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service