Skip to Content
Merck
All Photos(2)

Key Documents

808482

Sigma-Aldrich

Poly(D,L-lactide-co-glycolide)

lactide:glycolide 50:50, acid and hydroxy terminated, Mn 25000

Synonym(s):

HO-PLGA-COOH, PLGA, heterobifunctional PLGA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HO(C3H4O2)x(C2H2O2)yH
UNSPSC Code:
12162002

form

crystals

mol wt

Mn 25000

storage temp.

2-8°C

Application

Heterobifunctional degradable PLGA for conjugation, grafting and functionalization for drug delivery applications.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Saurabh Srivastava et al.
Journal of cancer research and therapeutics, 15(3), 480-490 (2019-06-07)
The aim of the study to develop surface modified targeted moiety α-tocopherol (α-t) encapsulated with 5-fluorouracil (5-FU)-poly-D, L-lactic-co-glycolic acid nanoparticles (PLGA NPs) toward the anticancer activity against oral squamous cell carcinoma (OSCC). 5-FU was conjugated with the polymer, PLGA by
Grafting of telechelic poly(lactic-co-glycolic acid) onto O2 plasma-treated polypropylene flakes.
Ren J, et al.
Journal of Applied Polymer Science, 121, 210-216 (2011)
Kun Wei et al.
International journal of pharmaceutics, 464(1-2), 225-233 (2014-01-28)
Di-block polymer of poly (lactic-co-glycolic acid)-poly (ethylene glycol) (PLGA-PEG) end-capped with capecitabine (CAP) at the hydrophobic domain and folate (FA) at hydrophilic domain were synthesized respectively. The products were extensively studied by nuclear magnetic resonance ((1)H NMR) and gel permeation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service