Skip to Content
Merck
All Photos(2)

Key Documents

772631

Sigma-Aldrich

Gallium

pieces, 99.99999% trace metals basis

Synonym(s):

Gallium element

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Ga
CAS Number:
Molecular Weight:
69.72
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.23
Pricing and availability is not currently available.

Assay

99.99999% trace metals basis

form

pieces
(metallic liquid or solid (ambient temp dependent))

resistivity

25.795 μΩ-cm, 30°C

bp

2403 °C (lit.)

mp

29.8 °C (lit.)

density

5.904 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

[Ga]

InChI

1S/Ga

InChI key

GYHNNYVSQQEPJS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Gallium metal is used in many high technology applications including the growth of gallium semiconductor nanoparticles by MBE[1] and as a catalyst for the formation of nanoscale semiconductors such as PbSe[2] .

Pictograms

CorrosionExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Met. Corr. 1

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jia Zhu et al.
Nano letters, 7(4), 1095-1099 (2007-03-14)
Lead chalcogenide nanostructures are good potential candidates for applications in multiexciton solar cells, infrared photodetectors, and electroluminescence devices. Here we report the synthesis and electrical measurements of hyperbranched PbSe nanowire networks. Hyperbranched PbSe nanowire networks are synthesized via a vapor-liquid-solid
Josef A Czaban et al.
Nano letters, 9(1), 148-154 (2009-01-16)
We report the use of Te as an n-type dopant in GaAs core-shell p-n junction nanowires for use in photovoltaic devices. Te produced significant change in the morphology of GaAs nanowires grown by the vapor-liquid-solid process in a molecular beam
Michael A Jakupec et al.
Metal ions in biological systems, 42, 425-462 (2004-06-23)
Gallium has been the second metal to show activity against malignant tumors in humans soon after the establishment of platinum drugs in routine clinical practice. It has the unique property of inhibiting tumor growth as a simple cation, mainly because
Prasana Sahoo et al.
Biosensors & bioelectronics, 44, 164-170 (2013-02-22)
We demonstrate a very simple and generic protocol for ultrasensitive in-situ label-free detection of DNA hybridization using third generation poly(amidoamine)dendrimer (G3-PAMAM) functionalized GaN nanowires (NWs). PAMAM modified GaN NWs provides large density of docking site to immobilize significant number of
Andrew B Kelson et al.
Current opinion in pharmacology, 13(5), 707-716 (2013-07-24)
Microbes have evolved elaborate iron-acquisition systems to sequester iron from the host environment using siderophores and heme uptake systems. Gallium(III) is structurally similar to iron(III), except that it cannot be reduced under physiological conditions, therefore gallium has the potential to

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service