Skip to Content
Merck
All Photos(1)

Key Documents

755753

Sigma-Aldrich

Chromium(0) hexacarbonyl

packaged for use in deposition systems

Synonym(s):

Chromium(0) hexacarbonyl, Chromiumhexacarbonyl, Hexacarbonylchromium(0)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cr(CO)6
CAS Number:
Molecular Weight:
220.06
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

vapor density

7.6 (vs air)

vapor pressure

1 mmHg ( 36 °C)

form

powder, crystals or chunks

reaction suitability

core: chromium

mp

>150 °C (dec.) (lit.)

density

1.77 g/mL at 25 °C (lit.)

SMILES string

[Cr].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+]

InChI

1S/6CO.Cr/c6*1-2;

InChI key

KOTQLLUQLXWWDK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Chromiumhexacarbonyl is a volatile; air stable precursor of Chromium(0); widely used for thin film deposition - ALD and CVD. The thin films can be grown at room temperature and low pressure by laser CVD .

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wang, S-C.; Lin H-T.; Nayak, P. K.; Chang, S-Y.; Huang, J-L.
Thin Solid Films, 518, 7360-7360 (2010)
Maruyama, T.; Akagi, H.;
Journal of the Electrochemical Society, 143, 1955-1955 (1996)
Cheng, R.; Borca, C. N.; Dowben, P. A.; Stadler, S,; Idzerda, Y. U.
Applied Physics Letters, 78, 521-521 (2001)
Sousa, P. M.; Silvestre, A. J.; Conde, O.
Thin Solid Films, 519, 3653-3653 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service