Skip to Content
Merck
All Photos(1)

Key Documents

744042

Sigma-Aldrich

1-Butene

≥99.6%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2=CHCH2CH3
CAS Number:
Molecular Weight:
56.11
Beilstein:
1098262
EC Number:
MDL number:
UNSPSC Code:
12142100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

vapor density

1.93 (vs air)

vapor pressure

1939 mmHg ( 21.1 °C)

Assay

≥99.6%

form

gas

autoignition temp.

723 °F

expl. lim.

9.3 %

bp

−6.3 °C (lit.)

mp

−185 °C (lit.)

SMILES string

CCC=C

InChI

1S/C4H8/c1-3-4-2/h3H,1,4H2,2H3

InChI key

VXNZUUAINFGPBY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

500ml high pressure aluminum cylinder equipped with a stainless steel valve according to DIN 477-1.

Other Notes

For 744042-265G-EU: outlet fitting: DIN 477-1 no. 1, Z742160 or Z742161 are the recommended regulators.

Recommended products

For 744042-265G-EU: outlet fitting: DIN 477-1 no. 1, Z742160 or Z742126 are the recommended regulators.

Pictograms

FlameGas cylinder

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Flam. Gas 1A - Press. Gas Liquefied gas

Storage Class Code

2A - Gases

WGK

nwg

Flash Point(F)

-112.0 °F - closed cup

Flash Point(C)

-80 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Felicia R Lucci et al.
Nature communications, 6, 8550-8550 (2015-10-10)
Platinum is ubiquitous in the production sectors of chemicals and fuels; however, its scarcity in nature and high price will limit future proliferation of platinum-catalysed reactions. One promising approach to conserve platinum involves understanding the smallest number of platinum atoms
Kazuhiro Taguchi et al.
SpringerPlus, 3, 6-6 (2015-02-13)
An estimation procedure for biobased carbon content of polyethylene composite was studied using carbon-14 ((14)C) concentration ratios as measured by accelerated mass spectrometry (AMS). Prior to the measurement, additives and fillers in composites should be removed because they often contain
A Stojić et al.
Environmental science and pollution research international, 22(17), 13137-13152 (2015-05-01)
In this study, the concentrations of volatile organic compounds were measured by the use of proton transfer reaction mass spectrometry, together with NO x , NO, NO2, SO2, CO and PM10 and meteorological parameters in an urban area of Belgrade

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service