Skip to Content
Merck
All Photos(1)

Key Documents

736120

Supelco

α,α,α-Trifluorotoluene solution

NMR reference standard, 0.05% in benzene-d6 (99.6 atom % D), NMR tube size 5 mm × 8 in., NMR tube wall thickness 0.24 mm , ultra-thin wall

Synonym(s):

Benzotrifluoride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CF3
CAS Number:
Molecular Weight:
146.11
EC Number:
MDL number:
UNSPSC Code:
12142201
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

NMR reference standard
analytical standard

concentration

0.05% in benzene-d6 (99.6 atom % D)

technique(s)

NMR: suitable

NMR tube size

5 mm × 8 in.

NMR tube wall thickness

0.24 mm , ultra-thin wall

suitability

suitable for NMR (reference standard)

format

single component solution

SMILES string

FC(F)(F)c1ccccc1

InChI

1S/C7H5F3/c8-7(9,10)6-4-2-1-3-5-6/h1-5H

InChI key

GETTZEONDQJALK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Features and Benefits

19F sensitivity

Quantity

5 mm O.D. tube contains 0.700 mL.

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 3 - Asp. Tox. 1 - Carc. 1A - Eye Irrit. 2 - Flam. Liq. 2 - Muta. 1B - Skin Irrit. 2 - STOT RE 1

Target Organs

Blood

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

12.2 °F

Flash Point(C)

-11 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shota Tsukada et al.
Neuropharmacology, 48(4), 479-491 (2005-03-10)
Glutamate transporters rapidly take up synaptically released glutamate and maintain the glutamate concentration in the synaptic cleft at a low level. (2S, 3S)-3-[3-[4-(trifluoromethyl)benzoylamino]benzyloxy]aspartate (TFB-TBOA) is a novel glutamate transporter blocker that potently suppresses the activity of glial transporters. TFB-TBOA inhibited
[Hygienic standardization of benzotrifluoride level in reservoir water].
V I Antonova et al.
Gigiena i sanitariia, (2)(2), 86-87 (1985-02-01)
T Thriveni et al.
Environmental monitoring and assessment, 151(1-4), 9-18 (2008-04-04)
An electroanalytical method has been developed for the determination of the herbicides ethalfluralin[N-ethyl-N-(2-methyl-2-propenyl)-2,6-dinitro-4-(trifluoromethyl) bezenamine] and methalpropalin [N-(2-methyl-2-propenyl)-2, 6-dinitro-N-propyl-4 (trifluoromethyl) benzenamine] by differential pulse adsorptive stripping voltammetry (DP-AdSV) on a hanging mercury drop electrode (HMDE) with universal buffer as supporting electrolyte.
R A Yadav et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 64(2), 454-463 (2006-01-18)
Force field calculations have been carried out for 2-amino-5-chloro- and 2-amino-5-bromo-benzotrifluorides using the earlier (Ref. of this article) reported IR and Raman spectra. As the frequencies of the corresponding modes for the C-Br and the C-Cl groups do not differ
C Yost et al.
Applied occupational and environmental hygiene, 15(1), 165-170 (2000-03-11)
Three solvents (OXSOL 10, monochlorotoluene or mixed isomers of 1- chloro-2-methyl benzene and 1-chloro-4-methyl benzene; OXSOL 100, parachlorobenzotrifluoride or 1-chloro-4-(trifluoromethyl) benzene; and OXSOL 2000, benzotrifluoride or trifluoromethyl benzene) produced by Occidental Chemical Corporation (Niagara Falls, NY) were considered as candidates

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service