Skip to Content
Merck
All Photos(2)

Key Documents

697281

Sigma-Aldrich

Tris(tert-butoxy)silanol

packaged for use in deposition systems

Synonym(s):

TBS

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
((CH3)3CO)3SiOH
CAS Number:
Molecular Weight:
264.43
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23
Pricing and availability is not currently available.

Assay

≥99.0% (GC)
≥99.998% trace metals basis (ICP)

form

solid

bp

205-210 °C (lit.)

mp

63-65 °C (lit.)

SMILES string

CC(C)(C)O[Si](O)(OC(C)(C)C)OC(C)(C)C

InChI

1S/C12H28O4Si/c1-10(2,3)14-17(13,15-11(4,5)6)16-12(7,8)9/h13H,1-9H3

InChI key

HLDBBQREZCVBMA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Atomic number of base material: 14 Silicon

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jonathan Stewart et al.
Modern pathology : an official journal of the United States and Canadian Academy of Pathology, Inc, 28(3), 428-436 (2014-09-27)
The oncogenic role of WNT is well characterized. Wntless (WLS) (also known as GPR177, or Evi), a key modulator of WNT protein secretion, was recently found to be highly overexpressed in malignant astrocytomas. We hypothesized that this molecule may be
Pedro A Dionísio et al.
Neurobiology of aging, 36(1), 228-240 (2014-12-03)
Alzheimer's disease (AD) is a neurodegenerative disorder hallmarked by the accumulation of extracellular amyloid-β (Aβ) peptide and intraneuronal hyperphosphorylated tau, as well as chronic neuroinflammation. Tauroursodeoxycholic acid (TUDCA) is an endogenous anti-apoptotic bile acid with potent neuroprotective properties in several
Sylvain Ladoire et al.
Autophagy, 11(10), 1878-1890 (2015-10-29)
In spite of adjuvant chemotherapy, a significant fraction of patients with localized breast cancer (BC) relapse after optimal treatment. We determined the occurrence of cytoplasmic MAP1LC3B/LC3B (microtubule-associated protein 1 light chain 3B)-positive puncta, as well as the presence of nuclear
Ádám I Csincsi et al.
Journal of immunology (Baltimore, Md. : 1950), 194(10), 4963-4973 (2015-04-10)
The physiological roles of the factor H (FH)-related proteins are controversial and poorly understood. Based on genetic studies, FH-related protein 5 (CFHR5) is implicated in glomerular diseases, such as atypical hemolytic uremic syndrome, dense deposit disease, and CFHR5 nephropathy. CFHR5
Paula Duarte-Guterman et al.
PloS one, 10(6), e0129880-e0129880 (2015-06-16)
Estradiol increases cell proliferation in the dentate gyrus of the female rodent but it is not known whether the G protein-coupled estrogen receptor (GPER), a membrane receptor, is involved in this process, nor whether there are regional differences in estradiol's

Articles

Atomic Layer Deposition (ALD) technology ensures uniform coating on complex 3D surfaces with precise chemisorption cycles.

Atomic Layer Deposition (ALD) technology ensures uniform coating on complex 3D surfaces with precise chemisorption cycles.

Atomic Layer Deposition (ALD) technology ensures uniform coating on complex 3D surfaces with precise chemisorption cycles.

Atomic Layer Deposition (ALD) technology ensures uniform coating on complex 3D surfaces with precise chemisorption cycles.

See All

Questions

  1. What is the Department of Transportation shipping information for this product?

    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service