Skip to Content
Merck
All Photos(1)

Key Documents

537268

Sigma-Aldrich

6-Methoxy-2-naphthoic acid

97%

Synonym(s):

6-Methoxy-2-naphthalenecarboxylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OC10H6CO2H
CAS Number:
Molecular Weight:
202.21
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Pricing and availability is not currently available.

Assay

97%

mp

201-206 °C (lit.)

functional group

carboxylic acid

SMILES string

COc1ccc2cc(ccc2c1)C(O)=O

InChI

1S/C12H10O3/c1-15-11-5-4-8-6-10(12(13)14)3-2-9(8)7-11/h2-7H,1H3,(H,13,14)

InChI key

YZBILXXOZFORFE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Used for preparation of arylamide inhibitors (HIV-1 integrase)

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Amaj Ahmed Laskar et al.
International journal of biological macromolecules, 96, 798-806 (2017-01-07)
Human salivary aldehyde dehydrogenase (HsALDH) appears to be the first line of defence against toxic aldehydes contained in exogenous sources and is important for maintaining healthy oral cavity and protection from oral cancer. Here, the activity and stability of purified

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service