About This Item
Recommended Products
vapor density
4.2 (vs air)
vapor pressure
0.2 mmHg ( 25 °C)
Assay
98%
form
liquid
expl. lim.
9.5 %
refractive index
n20/D 1.554 (lit.)
bp
205 °C (lit.)
mp
−63 °C (lit.)
density
0.963 g/mL at 25 °C (lit.)
functional group
amine
SMILES string
CCNc1ccccc1
InChI
1S/C8H11N/c1-2-9-8-6-4-3-5-7-8/h3-7,9H,2H2,1H3
InChI key
OJGMBLNIHDZDGS-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Application
It may be used in the synthesis of the following:
- 2-(N-ethylphenylamino)-1,4-benzoquinone[9]
- 2-(arylaminomethyl)phenylboronic acid[10]
- poly(methyl methacrylate) (PMMA) films containing NEA[11]
- poly(N-ethylaniline) (PNEA)[12]
- N-ethyl-N-phenylpicolinamide[13]
- 4-chloro-N-ethyl-N-phenylpicolinamide[13]
- N2,N6-diethyl-N2,N6-diphenylpyridine-2,6-dicarboxamide[13]
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - STOT RE 2
Storage Class Code
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
WGK
WGK 1
Flash Point(F)
185.0 °F - closed cup
Flash Point(C)
85 °C - closed cup
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service