Skip to Content
Merck
All Photos(1)

Key Documents

382159

Sigma-Aldrich

trans-3,5-Dimethoxy-4-hydroxycinnamaldehyde

98%

Synonym(s):

Sinapinaldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC6H2(OCH3)2CH=CHCHO
CAS Number:
Molecular Weight:
208.21
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Pricing and availability is not currently available.

Assay

98%

form

solid

mp

104-106 °C (lit.)

storage temp.

2-8°C

SMILES string

[H]C(=O)\C([H])=C(/[H])c1cc(OC)c(O)c(OC)c1

InChI

1S/C11H12O4/c1-14-9-6-8(4-3-5-12)7-10(15-2)11(9)13/h3-7,13H,1-2H3/b4-3+

InChI key

CDICDSOGTRCHMG-ONEGZZNKSA-N

Looking for similar products? Visit Product Comparison Guide

Application

trans-3,5-Dimethoxy-4-hydroxycinnamaldehyde (Sinapinaldehyde) is suitable for use in the determination of phenolic compounds by HPLC with multiple wavelength detector in grape marc distillates aged in Quercus petraea, Quercus robur and Quercus alba wooden barrels.[1] It is suitable for use in the GC/MS positive ion chemical ionization (PICI) characterization of volatile phenols and benzene aldehydes in various woods.[2]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mónica Schwarz et al.
Foods (Basel, Switzerland), 9(3) (2020-03-07)
The wine spirits used for the elaboration of Brandy de Jerez are mainly obtained from wines produced from the Airén type of grape, which comes from the vineyards located in the region of La Mancha (Central Spain). This entails a
David Fournand et al.
Phytochemistry, 62(2), 139-146 (2002-12-17)
Capillary zone electrophoresis has been used to monitor the first steps of the dehydrogenative polymerization of coniferyl alcohol, sinapyl aldehyde, or a mixture of both, catalyzed by the horseradish peroxidase (HRP)-H(2)O(2) system. When coniferyl alcohol was the unique HRP substrate
M H Farah et al.
Planta medica, 58(1), 14-18 (1992-02-01)
Coniferaldehyde, scopoletin, sinapaldehyde, and syringaldehyde were isolated from an aqueous extract of Senra incana. All four compounds inhibited prostaglandin synthetase in a dose-dependent way. Compared to aspirin, the potency of coniferaldehyde and scopoletin was about five times higher, whereas syringaldehyde
First Approach to the Analytical Characterization of Barrel-Aged Grape Marc Distillates Using Phenolic Compounds and Colour Parameters.
Rodriguez-Solana R, et al.
Food Technology and Biotechnology, 52(4), 391-402 (2014)
Erin K Bomati et al.
The Plant cell, 17(5), 1598-1611 (2005-04-15)
We describe the three-dimensional structure of sinapyl alcohol dehydrogenase (SAD) from Populus tremuloides (aspen), a member of the NADP(H)-dependent dehydrogenase family that catalyzes the last reductive step in the formation of monolignols. The active site topology revealed by the crystal

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service