Skip to Content
Merck
All Photos(1)

Key Documents

366455

Sigma-Aldrich

Triphenylphosphine, polymer-bound

100-200 mesh, extent of labeling: ~3.0 mmol/g loading, 2 % cross-linked with divinylbenzene

Synonym(s):

Copolymer of styrene and divinylbenzene, diphenylphosphinated, Diphenylphosphino-polystyrene, Polystyrene crosslinked with divinylbenzene, diphenylphosphinated

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
-C6H4P(C6H5)2
CAS Number:
Molecular Weight:
262.29
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

crosslinking

2 % cross-linked with divinylbenzene

reaction suitability

reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reaction type: solution phase peptide synthesis
reagent type: ligand

extent of labeling

~3.0 mmol/g loading

particle size

100-200 mesh

functional group

phosphine

SMILES string

c1ccc(cc1)P(c2ccccc2)c3ccccc3

InChI

1S/C18H15P/c1-4-10-16(11-5-1)19(17-12-6-2-7-13-17)18-14-8-3-9-15-18/h1-15H

InChI key

RIOQSEWOXXDEQQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Preparation Note

Used for the preparation of supported precious metal complexes.[1]
Used to prepare polymer bound ylides for Wittig reactions. It can also be used to convert alcohols or carboxylic acids to the corresponding chlorides under relatively mild conditions in high yield.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Clark, H.C., et al.
Journal of Organometallic Chemistry, 322, 393-393 (1987)
Uday Narayan Maiti et al.
ACS applied materials & interfaces, 7(46), 25898-25905 (2015-11-03)
A scalable and controllable nanoscale perforation method for graphene is developed on the basis of the two-step thermal activation of a graphene aerogel. Different resistance to the thermal oxidation between graphitic and defective domains in the weakly reduced graphene oxide
Crispin Lichtenberg et al.
Dalton transactions (Cambridge, England : 2003), 44(46), 20056-20066 (2015-11-04)
A diolefin ether, trop2O (2), and a diolefin thioether, trop2S (3), have been investigated as ligand analogues of the well-established diolefin amine, trop2NH (1). Compounds 2 and 3 form different conformers in solution and in the solid state. Whereas 2
Hui Yang et al.
Toxicology letters, 230(1), 10-18 (2014-08-08)
Gold nanoparticles (GNPs) have considerable applications in biomedicine, such as in bio-sensing, bio-imaging, drug delivery and photothermal therapeutics. However, currently there are limited information regarding the impact of pregnancy on their biodistribution, elimination and toxicity. In this study, we investigated
Seyed Mohammad Davachi et al.
Materials science & engineering. C, Materials for biological applications, 58, 294-309 (2015-10-20)
In this study, the encapsulated triclosan with a low molecular weight PLLA (LATC30) is dispersed into a PLLA having higher molecular weight via melt blending to increase the overall properties and particularly antibacterial activity of the system. The proposed method

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service