Skip to Content
Merck
All Photos(1)

Key Documents

345016

Sigma-Aldrich

2-Methyl-5-nitrobenzonitrile

97%

Synonym(s):

5-Nitro-o-tolunitrile

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
O2NC6H3(CH3)CN
CAS Number:
Molecular Weight:
162.15
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

97%

mp

103.5-107.5 °C (lit.)

solubility

ethanol: soluble(lit.)

functional group

nitrile
nitro

SMILES string

Cc1ccc(cc1C#N)[N+]([O-])=O

InChI

1S/C8H6N2O2/c1-6-2-3-8(10(11)12)4-7(6)5-9/h2-4H,1H3

InChI key

XOSDYLFXPMFRGF-UHFFFAOYSA-N

General description

2-Methyl-5-nitrobenzonitrile is an aralkyl nitrile.[1] It is formed by nitration of o-toluonitrile with NO2+BF4-.[1] Crystal structure of 2-methyl-5-nitrobenzonitrile is stabilized by van der Waals interactions.[2]

Application

2-Methyl-5-nitrobenzonitrile was used in the preparation of 5-(2-methyl-5-nitrophenyl)-1H-tetrazole.[3]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Recent aspects of nitration: New preparative methods and mechanistic studies (A Review).
Olah GA, et al.
Proceedings of the National Academy of Sciences of the USA, 79(14), 4487-4494 (1982)
Jing Dai et al.
Acta crystallographica. Section E, Structure reports online, 64(Pt 10), o1944-o1944 (2008-01-01)
In the title compound, C(8)H(7)N(5)O(2), the benzene ring makes a dihedral angle of 45.7 (2)° with the tetra-zole ring. In the crystal structure, the mol-ecules are linked into a chain running along the a axis by N-H⋯N hydrogen bonds, and the
Wen-Xian Liang et al.
Acta crystallographica. Section E, Structure reports online, 64(Pt 6), o972-o972 (2008-01-01)
In the title compound, C(8)H(6)N(2)O(2), the nitro group is rotated by 10.2 (2)° out of the plane of the benzene ring. The crystal structure is stabilized by van der Waals inter-actions.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service