Skip to Content
Merck
All Photos(1)

Key Documents

293512

Sigma-Aldrich

7-Hydroxy-3,4,8-trimethylcoumarin

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H12O3
CAS Number:
Molecular Weight:
204.22
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

97%

mp

278-280 °C (lit.)

functional group

ester

SMILES string

CC1=C(C)c2ccc(O)c(C)c2OC1=O

InChI

1S/C12H12O3/c1-6-7(2)12(14)15-11-8(3)10(13)5-4-9(6)11/h4-5,13H,1-3H3

InChI key

GNBLUSRSAGXTJN-UHFFFAOYSA-N

General description

7-Hydroxy-3,4,8-trimethylcoumarin is an inhibitor of melanin production[1].

Application

7-Hydroxy-3,4,8-trimethylcoumarin has been used:
  • to investigate the effect of melanin inhibiting compounds on mycelial pigmentation of Aspergillus bridgeri grown on potato dextrose agar[1]
  • in the preparation of 7-ethynyl-3,4,8-trimethylcoumarin[2]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jiawang Liu et al.
Chemical research in toxicology, 25(5), 1047-1057 (2012-03-27)
To discover new selective mechanism-based P450 inhibitors, eight 7-ethynylcoumarin derivatives were prepared through a facile two-step synthetic route. Cytochrome P450 activity assays indicated that introduction of functional groups in the backbone of coumarin could enhance the inhibition activities toward P450s
C Ganesh Kumar et al.
Letters in applied microbiology, 53(3), 350-358 (2011-07-06)
The aim of the study is to isolate and characterize a melanin pigment from a new strain of Aspergillus bridgeri isolated from rhizosphere soil of Eucalyptus tree and to investigate its antioxidant activity. The extracellular pigment was alkali soluble, acid-resistant

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service