Skip to Content
Merck
All Photos(1)

Key Documents

264733

Sigma-Aldrich

4-Hexen-3-one, predominantly trans

≥90%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH=CHCOC2H5
CAS Number:
Molecular Weight:
98.14
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

≥90%

refractive index

n20/D 1.44 (lit.)

bp

135-137 °C (lit.)

density

0.858 g/mL at 25 °C (lit.)

SMILES string

CCC(=O)\C=C\C

InChI

1S/C6H10O/c1-3-5-6(7)4-2/h3,5H,4H2,1-2H3/b5-3+

InChI key

FEWIGMWODIRUJM-HWKANZROSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Kinetcis of the reactions of hydroxyl radicals and chlorine atoms with 4-hexen-3-one has been investigated.

Application

4-Hexen-3-one has been used to obtain oxidation reference spectra to monitor the oxidation of edible oils by FTIR analysis.

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

93.2 °F - closed cup

Flash Point(C)

34 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Monitoring the oxidation of edible oils by Fourier transform infrared spectroscopy.
Van de Voort FR, et al.
Journal of the American Oil Chemists' Society, 71(3), 243-253 (1994)
María B Blanco et al.
The journal of physical chemistry. A, 116(24), 6033-6040 (2012-01-26)
Rate coefficients for the reactions of hydroxyl radicals and chlorine atoms with 4-hexen-3-one, 5-hexen-2-one, and 3-penten-2-one have been determined at 298 ± 2 K and atmospheric pressure of air. Rate coefficients for the compounds were determined using a relative kinetic
Guangwei Si et al.
Neuron, 101(5), 950-962 (2019-01-27)
Odor perception allows animals to distinguish odors, recognize the same odor across concentrations, and determine concentration changes. How the activity patterns of primary olfactory receptor neurons (ORNs), at the individual and population levels, facilitate distinguishing these functions remains poorly understood. Here, we

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service