Skip to Content
Merck
All Photos(1)

Key Documents

262854

Sigma-Aldrich

Tantalum

rod, diam. 6.35 mm, ≥99.9% trace metals basis

Synonym(s):

Ta

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Ta
CAS Number:
Molecular Weight:
180.95
EC Number:
MDL number:
UNSPSC Code:
12141741
PubChem Substance ID:
NACRES:
NA.23

vapor pressure

<0.01 mmHg ( 537.2 °C)

Assay

≥99.9% trace metals basis

form

rod

autoignition temp.

572 °F

resistivity

13.5 μΩ-cm, 20°C

diam.

6.35 mm

bp

5425 °C (lit.)

mp

2996 °C (lit.)

density

16.69 g/cm3 (lit.)

SMILES string

[Ta]

InChI

1S/Ta

InChI key

GUVRBAGPIYLISA-UHFFFAOYSA-N

Quantity

25 g = 5 cm; 125 g = 25 cm

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Scott M Sporer et al.
Clinical orthopaedics and related research, 470(11), 3156-3163 (2012-09-25)
Stabilization of a pelvic discontinuity with a posterior column plate with or without an associated acetabular cage sometimes results in persistent micromotion across the discontinuity with late fatigue failure and component loosening. Acetabular distraction offers an alternative technique for reconstruction
Orbital ultrasound in the selective screening of a dural-cavernous sinus fistula.
Hamid Shokoohi et al.
European journal of emergency medicine : official journal of the European Society for Emergency Medicine, 20(2), 137-139 (2013-02-26)
Hany A El-Sayed et al.
Nanoscale, 5(4), 1494-1498 (2013-01-23)
Ta oxide nanotubes (NTs) were formed by the anodization of Ta at 15 V in a solution of concentrated sulfuric acid containing 0.8-1.0 M hydrofluoric acid. To study the initial stages of NT formation, FESEM images of samples anodized for
Thomas Larzon et al.
Vascular, 21(2), 87-91 (2013-03-20)
Control of back bleeding from the hypogastric artery into the aneurysm after endovascular aneurysm repair (EVAR) of a ruptured aorto-iliac aneurysm may be necessary in order to avoid a type II endoleak. It is an emergency situation and selective catheterization
Matthew D Budge et al.
Journal of shoulder and elbow surgery, 22(4), 535-541 (2012-09-11)
Aseptic loosening of all-polyethylene glenoid components remains a limiting factor in achieving long-term implant survival in total shoulder arthroplasty (TSA). This study prospectively evaluated the functional and radiographic outcomes of patients undergoing TSA with a novel, porous, tantalum-backed glenoid component

Articles

Biomedical implants are essentially foreign substances within the human body that must survive many years’ exposure to demanding mechanical and physiological conditions. Despite these challenges, metal implants have been widely used to substitute for or rebuild hard tissues such as bones and teeth.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service