261300
Ytterbium
ingot, 99.9% trace rare earth metals basis
About This Item
Recommended Products
Assay
99.9% trace rare earth metals basis
form
ingot
reaction suitability
reagent type: catalyst
core: ytterbium
resistivity
28 μΩ-cm, 20°C
bp
1196 °C (lit.)
mp
824 °C (lit.)
density
6.54 g/mL at 25 °C (lit.)
SMILES string
[Yb]
InChI
1S/Yb
InChI key
NAWDYIZEMPQZHO-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
A significant limiting factor for wearable electronics and wireless sensors is the finite amount of energy that can be stored in on-board batteries.
Rechargeable solid-state batteries are becoming increasingly important due to wide-spread use in computers, portable electronics, and vehicular applications.
Rechargeable solid-state batteries are becoming increasingly important due to wide-spread use in computers, portable electronics, and vehicular applications.
Rechargeable solid-state batteries are becoming increasingly important due to wide-spread use in computers, portable electronics, and vehicular applications.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service