Skip to Content
Merck
All Photos(1)

Key Documents

205842

Sigma-Aldrich

Palladium(II) oxide hydrate

Pd content ~97 %

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
OPd · xH2O
CAS Number:
Molecular Weight:
122.42 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.23
Pricing and availability is not currently available.

form

powder and chunks

composition

Degree of hydration, ~1
Pd content, ~97%

reaction suitability

reagent type: catalyst
core: palladium

SMILES string

O=[Pd].[H]O[H]

InChI

1S/H2O.O.Pd/h1H2;;

InChI key

CJCUJYURPLYQOL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

A new AB2O4 phase, LuPd2O4, with potential uses in superconducting material as well as being of geochemical importance, was formed at elevated pressure and temperature from Lu2O3 and PdO in the presence of potassium chlorate.[1]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chen, B-H.; Walker, D.
Chemistry of Materials, 9, 1700-1700 (1997)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service