Skip to Content
Merck
All Photos(1)

Key Documents

141054

Sigma-Aldrich

2-Amino-4-phenyl-5-tetradecylthiazole

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H36N2S
CAS Number:
Molecular Weight:
372.61
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

mp

72-74 °C (lit.)

functional group

phenyl

SMILES string

CCCCCCCCCCCCCCc1sc(N)nc1-c2ccccc2

InChI

1S/C23H36N2S/c1-2-3-4-5-6-7-8-9-10-11-12-16-19-21-22(25-23(24)26-21)20-17-14-13-15-18-20/h13-15,17-18H,2-12,16,19H2,1H3,(H2,24,25)

InChI key

WYDPIVMKULHMBF-UHFFFAOYSA-N

General description

2-Amino-4-phenyl-5-tetradecylthiazole acts as ligand for functionalizing the silica-based adsorbents.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Heavy Metallic and Organometallic Ions Scavenging Using Silica-Based Adsorbent Functionalized with Ligands Containing Sulfur and Nitrogen Elements.
Yeh J-D and Chen S.
J. Chin. Chem. Soc., 59(1), 98-106 (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service