MilliporeSigma
  • Conditional Müllercell ablation causes independent neuronal and vascular pathologies in a novel transgenic model.
Conditional Müllercell ablation causes independent neuronal and vascular pathologies in a novel transgenic model.
The Journal of neuroscience : the official journal of the Society for Neuroscience (2012-11-09)
Weiyong Shen, Marcus Fruttiger, Ling Zhu, Sook H Chung, Nigel L Barnett, Joshua K Kirk, SoRa Lee, Nathan J Coorey, Murray Killingsworth, Larry S Sherman, Mark C Gillies
ABSTRACT
Müller cells are the major glia of the retina that serve numerous functions essential to retinal homeostasis, yet the contribution of Müller glial dysfunction to retinal diseases remains largely unknown. We have developed a transgenic model using a portion of the regulatory region of the retinaldehyde binding protein 1 gene for conditional Müller cell ablation and the consequences of primary Müller cell dysfunction have been studied in adult mice. We found that selective ablation of Müller cells led to photoreceptor apoptosis, vascular telangiectasis, blood-retinal barrier breakdown and, later, intraretinal neovascularization. These changes were accompanied by impaired retinal function and an imbalance between vascular endothelial growth factor-A (VEGF-A) and pigment epithelium-derived factor. Intravitreal injection of ciliary neurotrophic factor inhibited photoreceptor injury but had no effect on the vasculopathy. Conversely, inhibition of VEGF-A activity attenuated vascular leak but did not protect photoreceptors. Our findings show that Müller glial deficiency may be an important upstream cause of retinal neuronal and vascular pathologies in retinal diseases. Combined neuroprotective and anti-angiogenic therapies may be required to treat Müller cell deficiency in retinal diseases and in other parts of the CNS associated with glial dysfunction.
Sigma-Aldrich
Ciliary Neurotrophic Factor human, CNTF, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture
Pricing and availability is not currently available.
Roche
In Situ Cell Death Detection Kit, Fluorescein, sufficient for ≤50 tests, suitable for detection
Pricing and availability is not currently available.