Merck
  • Apolipoprotein C-III: a potent modulator of hypertriglyceridemia and cardiovascular disease.
Apolipoprotein C-III: a potent modulator of hypertriglyceridemia and cardiovascular disease.
Current opinion in endocrinology, diabetes, and obesity (2015-02-19)
Alison B Kohan
ABSTRACT
The purpose of this article is to summarize the recent epidemiological, basic science, and pharmaceutical research linking apolipoprotein C-III (apoC-III) with the development and treatment of cardiovascular disease (CVD). ApoC-III is an important emerging target linking hypertriglyceridemia with CVD. ApoC-III is a potent modulator of many established CVD risk factors, and is found on chylomicrons, very-low density lipoprotein, low-density lipoprotein, and high-density lipoprotein particles. Recent studies show that in humans, apoC-III levels are an independent risk factor for CVD, and its presence on lipoproteins may promote their atherogenicity. This year, two large-scale epidemiological studies have linked mutations in apoC-III with increased incidence of CVD and hypertriglyceridemia. ApoC-III raises plasma triglycerides through inhibition of lipoprotein lipase, stimulation of very-low density lipoprotein secretion, and is a novel factor in modulating intestinal triglyceride trafficking. ApoC-III also stimulates inflammatory processes in the vasculature and the pancreas. The combination of raising plasma triglycerides and independently stimulating inflammatory processes makes apoC-III a valuable target for reducing the residual CVD risk in patients already on statin therapy, or for whom triglycerides are poorly controlled. Clinical trials on apoC-III antisense oligonucleotides are in progress. ApoC-III is a potent direct modulator of established CVD risk factors: plasma triglycerides and inflammation. Recent findings show that changes in apoC-III levels are directly associated with changes in cardiovascular risk and the atherogenicity of the lipoproteins on which apoC-III resides. Emerging roles of apoC-III include a role in directing the atherogenicity of high-density lipoprotein, intestinal dietary triglyceride trafficking, and modulating pancreatic β-cell survival. The combination of these roles makes apoC-III an important therapeutic target for the management and prevention of CVD.
Sigma-Aldrich
Apolipoprotein C-III, human