Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MABS1220

Sigma-Aldrich

Anti-LTP/CETP Antibody, clone 14–8F

clone 14-8F, from mouse

Synonym(s):

Cholesteryl ester transfer protein, CETP, Lipid transfer protein I, LTP

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

14-8F, monoclonal

species reactivity

mouse, human, rabbit

technique(s)

ELISA: suitable
western blot: suitable

isotype

IgG2aκ

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... CETP(1071)

General description

Cholesteryl ester transfer protein (UniProt P22687; also known as CETP, Lipid transfer protein I, LTP) is encoded by the CETP (also known as HDLCQ10) gene (Gene ID 1071) in Oryctolagus cuniculus (Rabbit) species. Cholesteryl ester transfer protein (CETP) or lipid transfer protein (LTP) mediates the transport/exhanges of triglycerides and cholesteryl esters (CEs) between very-low-density lipoproteins (VLDL)/low-density lipoproteins (LDL) and high-density lipoproteins (HDL) in both directions. CETP plays an important role in the reverse cholesterol transport (RCT) pathway that mediates the transport of cholesterol from peripheral cells and tissues to the liver for metabolism and excretion in the bile. CETP shuttles CEs from HDL particles to apolipoprotein B-/apoB-containing particles in exchange for triglycerides. CETP is thought to facilitate the generation of small, dense LDL (sd LDL) particles through an indirect mechanism of increased rate of triglyceride transfer from VLDL in exchange for CEs in LDL and HDL. CE transfer between lipoproteins plays a crucial role in intravascular LDL remodeling. Decreased levels of HDL cholesterol and elevated levels of LDL is a risk factor for atherosclerosis and coronary heart disease (CHD).

Specificity

Clone 14-8F (a.k.a. 14-8H) reacts with both non-reduced and reduced LTP/CETP. Unlike clone 3-11D (3-9F; Cat. No. MABS1219), clone 14-8F (14-8H) did not block the lipid association of LTP/CETP with microemulsion, while both clones effectively blocked LTP/CETP-mediated triglyceride (TG) transfer from lLDL to HDL and from HDL to to apoB-containing lipoproteins (Ko, K.W., et al. (1994). J. Biol. Chem. 269(45):28206-28213).

Immunogen

Epitope: C-terminal region.
Native rabbit plasma LTP.

Application

Detect LTP/CETP using this Anti-LTP/CETP Antibody, clone 14–8F validated for use in Western Blotting, ELISA, Inhibits Activity/Function.
ELISA Analysis: Representative lots were conjugated with HRP and used as the detection antibody in combination with clone 3-11D (Cat. No. MABS1219) as the capture antibody for the detection of human plasma LTP/CETP by sandwich ELISA (Tosheska, K., et al. (2011). Bosn. J. Basic. Med. Sci. 11(3):169–173; Saito, K., et al. J. Lipid Res. (1999). 40(11):2013-2021; Sasai, K., et al. (1998). Clin. Chem. 44(7):1466-1473).
ELISA Analysis: A representative lot was conjugated with HRP and used as the detection antibody in combination with clone 3-11D (Cat. No. MABS1219) as the capture antibody for the detection of mouse plasma LTP/CETP by sandwich ELISA (Wu, C.A., et al. (2002). Arterioscler .Thromb. Vasc. Biol. 22(8):1347-1353).
ELISA Analysis: A representative lot, when conjugated with HRP and used as the detection antibody, showed pH-dependent LTP/CETP-binding activity with the maximum reactivity at a weakly acidic condition of pH 5 to 5.5 in sandwich ELISA applications (Saito, K., et al. J. Lipid Res. (1999). 40(11):2013-2021).
Western Blotting Analysis: A representative lot detected both rabbit and human LTP/CETP by Western blotting under either reducing or non-reducing condition (Ko, K.W., et al. (1994). J. Biol. Chem. 269(45):28206-28213).
Inhibits Activity/Function: A representative lot inhibited both rabbit and human LTP/CETP-mediated triglyceride (TG), but not cholesteryl ester (CE), transfer from low density lipoprotein (LDL) to high density lipoprotein (HDL) (Ko, K.W., et al. (1994). J. Biol. Chem. 269(45):28206-28213).
Inhibits Activity/Function: A representative lot effectively inhibited human LTP/CETP-mediated triglyceride (TG) transfer from high density lipoprotein (HDL) to to apoB-containing lipoproteins in human plasma. Clone 14-8F (14-8H) also inhibited cholesteryl ester (CE) transfer in the same assay by 40% (Ko, K.W., et al. (1994). J. Biol. Chem. 269(45):28206-28213).
Research Category
Signaling
Research Sub Category
Lipid Metabolism & Weight Regulation

Quality

Evaluated by Western Blotting purified Lipid transfer protein (CETP).

Western Blotting Analysis: 0.5 µg/mL of this antibody detected purified human plasma LTP/CETP.

Target description

~58 kDa observed. Target band appears larger than the calculated molecular weight of 54.5 kDa due to glycosylation.

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG2aκ antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service