Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.01660

Sigma-Aldrich

Butyllithium

Synonym(s):

BuLi solution, 1-Lithiobutane, n-Butyllithium, NBL

Sign Into View Organizational & Contract Pricing


About This Item

vapor pressure

163 hPa ( 20 °C)

form

liquid

autoignition temp.

240 °C

expl. lim.

1.2-7.8 % (v/v)

bp

69 °C/1013 hPa

mp

-95 °C

transition temp

flash point -22 °C

density

0.68 g/cm3 at 20 °C

General description

15% solution in n-hexane for synthesis

Storage and Stability

Store below +30°C.

Analysis Note

Assay (acidimetric):14.0 - 16.0 %

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Eye Dam. 1 - Flam. Liq. 2 - Pyr. Liq. 1 - Repr. 2 - Skin Corr. 1B - STOT RE 1 Inhalation - STOT SE 3 - Water-react 1

Target Organs

Central nervous system, Nervous system

Supplementary Hazards

Storage Class Code

4.2 - Pyrophoric and self-heating hazardous materials

WGK

WGK 3

Flash Point(F)

-7.6 °F

Flash Point(C)

-22 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service