Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

218766-M

Millipore

Caspase-4 Inhibitor I, Cell-Permeable

The Caspase-4 Inhibitor I, Cell-Permeable controls the biological activity of Caspase-4. This small molecule/inhibitor is primarily used for Cancer applications.

Synonym(s):

Caspase-4 Inhibitor I, Cell-Permeable, LEVD-CHO, Cell-permeable; sequence

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C96H164N20O25
Molecular Weight:
1998.45
UNSPSC Code:
12352200

Quality Level

Assay

≥97% (HPLC)

form

lyophilized powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

solubility

DMSO: 5 mg/mL

storage temp.

−20°C

General description

A potent, cell-permeable, and reversible inhibitor of caspase-4. The N-terminal sequence (amino acid residues 1-16) corresponds to the hydrophobic region of the signal peptide of Kaposi fibroblast growth factor and confers cell permeability to the peptide.
Potent, cell-permeable and reversible inhibitor of caspase-4.

Biochem/physiol Actions

Primary Target
caspase-4

Warning

Toxicity: Standard Handling (A)

Sequence

Ac-Ala-Ala-Val-Ala-Leu-Leu-Pro-Ala-Val-Leu-Leu-Ala-Leu-Leu-Ala-Pro-Leu-Glu-Val-Asp-CHO

Reconstitution

Following reconstitution aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Thornberry, N.A., and Lazebnik, Y. 1998. Science 281, 1312.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service