Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

S-090

Supelco

UR-144 5-Pentanoic acid metabolite-D5 (indole-D5)

100 μg/mL in methanol, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H22D5NO3
Molecular Weight:
346.47
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

100 μg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology
forensics and toxicology

format

single component solution

storage temp.

−20°C

SMILES string

O=C(C1C(C)(C)C1(C)C)C2=C([2H])N(CCCCC(O)=O)C3=C([2H])C([2H])=C([2H])C([2H])=C32

InChI

1S/C21H27NO3/c1-20(2)19(21(20,3)4)18(25)15-13-22(12-8-7-11-17(23)24)16-10-6-5-9-14(15)16/h5-6,9-10,13,19H,7-8,11-12H2,1-4H3,(H,23,24)/i5D,6D,9D,10D,13D

InChI key

UUTHIAPDCFFQKQ-FOZLTFMXSA-N

General description

An internal standard suitable for quantitation of UR-144 5-pentanoic acid metabolite levels using LC-MS/MS or GC/MS for synthetic cannabinoid testing methods in forensic analysis, clinical toxicology, or urine drug testing. UR-144 5-pentanoic acid metabolite is a primary urinary metabolite of UR-144, a third generation synthetic cannabinoid that has seen wide popularity in illicit drug markets since late 2012. For those synthetic cannabinoids currently controlled at the federal level, our certified solution standards of these substances are DEA-exempt, allowing laboratories to place orders without additional regulatory paperwork.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service