Key Documents
CLS55640200
Pyrex® class A low-actinic volumetric flask, Pyrex® ST stopper
capacity 200 mL
Synonym(s):
flasks glass, volumetric flasks, volumetric glass
About This Item
UNSPSC Code:
41121804
NACRES:
NB.43
Joint
joint stopper13
manufacturer/tradename
Corning 55640-200
Corning 55640
tol. measuring range
0.10 mL
capacity
200 mL
General description
Low actinic stained glass provides protection for materials sensitive to light. The protective color is an integral part of flask, which retains mechanical strength, chemical stability, and thermal resistance of Pyrex brand labware.
Legal Information
PYREX is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)