TMO143761
Nunc® MicroWell 96-well Plate with high flange skirt
Nunclon® Delta surface, polystyrene, with lid, clear, case of 50 ea
Synonym(s):
multiwell plate, treated multiwell plate, treated plate
All Photos(1)
About This Item
UNSPSC Code:
41122102
NACRES:
NB.13
material:
clear
polystyrene
polystyrene
size:
96 well
Recommended Products
material
clear
polystyrene
packaging
case of 50 ea
manufacturer/tradename
Nalge-Nunc 143761
size
96 well
Related Categories
General description
Obtain consistent and reliable results with Thermo Scientific Nunc MicroWell 96-Well Microplates. With multiple surface modifications, these plates are ideal for cell-based assays as well as biochemical assays. The flat bottom shape is suitable for microscopic and optical measurements. Available in either the high flange skirt type or the low flange pinchbar design, the plates are optimized for a range of robotics, automated readers and liquid handling systems.
Legal Information
Nunc is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Nunclon is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Choose from one of the most recent versions:
Certificates of Analysis (COA)