PHR2988
Star Anise Oil
pharmaceutical secondary standard, certified reference material
Synonym(s):
Anise star oil
About This Item
grade
certified reference material
pharmaceutical secondary standard
Quality Level
Agency
traceable to USP 1036960
API family
anise oil
optical activity
[α]20/D −1°, neat
CofA
current certificate can be downloaded
packaging
pkg of 1 g
refractive index
n20/D 1.554 (lit.)
bp
232 °C (lit.)
density
0.980 g/mL at 25 °C (lit.)
application(s)
pharmaceutical
storage temp.
2-8°C
Looking for similar products? Visit Product Comparison Guide
General description
Application
Analysis Note
Other Notes
related product
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Chronic 2 - Skin Sens. 1
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service