All Photos(1)
About This Item
Linear Formula:
HOCH213CH(NH2)13CO2H
CAS Number:
Molecular Weight:
107.08
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
Recommended Products
isotopic purity
99 atom % 13C
form
solid
optical activity
[α]25/D +14.6°, c = 2 in 1 M HCl
mp
222 °C (dec.) (lit.)
mass shift
M+2
SMILES string
N[13C@@H](CO)[13C](O)=O
InChI
1S/C3H7NO3/c4-2(1-5)3(6)7/h2,5H,1,4H2,(H,6,7)/t2-/m0/s1/i2+1,3+1
InChI key
MTCFGRXMJLQNBG-PIJHRLDMSA-N
Packaging
This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service