Skip to Content
Merck
All Photos(1)

Documents

129410

Sigma-Aldrich

2-Methyl-1-nonene

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)5CH2C(CH3)=CH2
CAS Number:
Molecular Weight:
140.27
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

refractive index

n20/D 1.424 (lit.)

bp

100 °C/68 mmHg (lit.)

density

0.745 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

CCCCCCCC(C)=C

InChI

1S/C10H20/c1-4-5-6-7-8-9-10(2)3/h2,4-9H2,1,3H3

InChI key

YLZQHQUVNZVGOK-UHFFFAOYSA-N

Application

2-Methyl-1-nonene is used to study the rate constant for the gas-phase reaction of NO(3) radicals and O(3) using relative rate methods.

Legal Information

Product of Rieke Metals, Inc.
Rieke is a registered trademark of Rieke Metals, Inc.

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

115.0 °F - closed cup

Flash Point(C)

46.1 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sherri A Mason et al.
The journal of physical chemistry. A, 113(19), 5649-5656 (2009-04-24)
Rate constants for the gas-phase reactions of NO(3) radicals and O(3) with a series of C(6)-C(14) 1-alkenes and 2-methyl-1-alkenes have been measured at 296 +/- 2 K and atmospheric pressure of air using relative rate methods. For the NO(3) radical

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service