Documentos clave
K5134-U
Omnifit® EZ Benchmark Columns with 2 Fixed Endpieces
I.D. × L 10 mm × 330 mm
About This Item
UNSPSC Code:
23151816
NACRES:
SB.52
manufacturer/tradename
Omnifit 006EZ-10-33-FF
Quality Level
technique(s)
LPLC: suitable
I.D. × L
10 mm × 330 mm
General description
Omnifit® Labware complete columns are supplied assembled and ready for use, with tubing and fittings to connect the column into an existing system. Two configurations are available:
  • Non-adjustable with two fixed endpieces (-FF), a cost-effective option when height adjustment is not important.
  • One fixed endpiece as a bed support and one adjustable endpiece (plunger) to adjust the bed height (-AF). The adjustable endpiece is designed to give the user fine control over the movement of the endpiece, minimizing the risk of disturbance to the packed bed and giving 80mm of height adjustment.
Legal Information
Omnifit is a registered trademark of Diba Industries
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)