Documentos clave
57174
Supelco
Portacolector ENVI-Disk
Process up to 6x 1L samples simultaneously
About This Item
UNSPSC Code:
12000000
Quality Level
technique(s)
solid phase extraction (SPE): suitable
application(s)
food and beverages
General description
The ENVI-Disk Holder Manifold holds one to six ENVI-Disk Holders with flasks, allowing you to simultaneously extract up to six 1-liter samples. Each of the six stations is controlled through an independent flow control valve. These valves are designed to vent the flask to the atmosphere when moved from the open to the closed position. The flow rate is controlled by the needle valve on the manifold.
The unit includes a sturdy polymer base with six stations, six flow control valves, a needle valve, a vacuum gauge, and vacuum tubing. A 1-liter glass bottle in the manifold acts as a trap, to protect the vacuum source in the event of an overflow from one of the sample flasks.
Legal Information
ENVI-Disk is a trademark of Sigma-Aldrich Co. LLC
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)