Documentos clave
HS15092D
Mini magnifer for use with mini led light box, free standing
About This Item
UNSPSC Code:
41121800
NACRES:
NB.51
manufacturer/tradename
Heathrow Scientific HS15092D
General description
Mini Magnifier enlarges objects for easier viewing . Swiveling, molded lens with 1.75X magnification. Dimensions: 6.3 x 5.3 x 8.1 in (16.1 x 13.5 x 20.7 cm). The perfect accessory for the eCount.
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)