Documentos clave
EP022265806
Eppendorf® Reagent Reservoir
pack of 10 ea
About This Item
UNSPSC Code:
41121522
material
polypropylene
packaging
pack of 10 ea
manufacturer/tradename
Eppendorf® 022265806
Categorías relacionadas
General description
Reagent reservoir Tip-Tub, autoclavable reservoir for the aspiration of liquids with multi-channel pipettes, 1 set = 10 reservoirs and 10 lids
Legal Information
Eppendorf is a registered trademark of Eppendorf AG