Skip to Content
Merck
All Photos(1)

Documents

MKL-1

09111801, human skin. Loose floating aggregates with irregular outline and no central necrosis.

Synonym(s):

MKL1 Cells

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

MKL-1, from human skin, 09111801

biological source

human skin

growth mode

Suspension

karyotype

Hyperdiploid

morphology

Loosely packed floating aggregates with irregular outline and no central necrosis

products

MKL-1 is positive for CM2B4/MCV T AE1/AE2, CK20, EMA, CAM5.2, NSE and TTF-1. MKL-1 is negative for CK7, chromagen, synaptophysin, neurofilament, LCA, vimentin and CD99.

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

metastasis

shipped in

dry ice

Cell Line Origin

Human neuroendocrine skin carcinoma

Cell Line Description

MKL-1 was established from a nodal metastasis in a 26-year-old Caucasian male patient. MKL-1 cells contain a Merkel cell polyomavirus integrated into the genome. MKL-1 cells cause tumour formation at 4-6 weeks at subcutaneous sites in athymic male nude mice (BALB/c nu/nu).

DNA Profile

STR-PCR Data:
Amelogenin: X,Y
CSF1PO: 11,12
D13S317: 8,11
D16S539: 10,12
D5S818: 11,12
D7S820: 8,11
THO1: 9,9.3
TPOX: 8
vWA: 16,18

Culture Medium

RPMI 1640 + 2 mM Glutamine + 10% FBS

Subculture Routine

Seed at 3 x 105 cells/ml in 5% CO2; 37 °C Saturation density is approximately 106 cells/ml. Population doubling time is around 120 hours.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service