Skip to Content
Merck
All Photos(1)

Documents

420360-M

Millipore

KMO Inhibitor I, Ro 61-8048

The KMO Inhibitor I, Ro 61-8048, also referenced under CAS 199666-03-0, controls the biological activity of KMO. This small molecule/inhibitor is primarily used for Neuroscience applications.

Synonym(s):

KMO Inhibitor I, Ro 61-8048, Kynurenine-3-Hydroxylase Inhibitor I, Ro 61-8048, 3,4-Dimethoxy-N-[4-(3-nitrophenyl)-2-thiazolyl]-benzenesulfonamide, Kynurenine-3-Monooxygenase Inhibitor I, Ro 61-8048, Dimethoxy-nitrophenyl-thiazolyl-benzenesulfonamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H15N3O6S2
CAS Number:
Molecular Weight:
421.45
MDL number:
UNSPSC Code:
12352200

Quality Level

Assay

≥99% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

yellow

solubility

DMSO: 100 mg/mL
ethanol: 12.5 mg/mL

storage temp.

2-8°C

InChI

1S/C17H15N3O6S2/c1-25-15-7-6-13(9-16(15)26-2)28(23,24)19-17-18-14(10-27-17)11-4-3-5-12(8-11)20(21)22/h3-10H,1-2H3,(H,18,19)

InChI key

NDPBMCKQJOZAQX-UHFFFAOYSA-N

General description

A cell-permeable thiazolylbenzenesulfonamide compound that acts as a potent, competitive and reversible inhibitor of kynurenine-3-monooxygenase activity (Ki = 4.8 nM; IC50 = 37 nM). Reported to increase the circulating levels of kynurenic acid (KYNA), an excitatory amino acid receptors antagonist, lower reactive oxygen species [ROS], reduce neuronal loss and enhance the survival rate in several neuronal disease models. An orally bioavilable KMO Inhibitor II, JM6, a prodrug of Ro 61-8048 is also available (Cat. No. 420361).

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Campesan, S., et al. 2011. Chem. Biol.21, 961.
Zwilling, D., et al. 2011. Cell145, 863.
Giorgini, F., et al. 2005. Nat. Genet.37, 526.
Cozzi, A.,. et al. 1999. J. Cereb. Blood Flow Metab.19, 771,
Rover, S., et al. 1997. J. Med. Chem.40, 4378.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service