Skip to Content
Merck
All Photos(1)

Documents

1.03085

Supelco

4-Chlorophenol solution

for AOX determination acc. to DIN EN ISO 9562 725 mg/l

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352104
NACRES:
NA.21

Quality Level

form

liquid

analyte functional class(es)

adsorbable organic halides (AOX)

specific analyte(s)

ammonium

technique(s)

photometry: suitable

pH

5 (20 °C in H2O)

density

1.00 g/cm3 at 20 °C

storage temp.

15-25°C

Application


  • Structure-activity relationships of aniline-based squaraines for distinguishable staining and bright two-photon fluorescence bioimaging in plant cells: Highlighting the advanced applications of Aniline in material science, this research demonstrates its use in developing squaraine dyes for cellular imaging, contributing significantly to cellular biology and diagnostics (Zhang et al., 2024).

  • Preparation of Two-Dimensional Polyaniline Sheets with High Crystallinity: This study exemplifies Aniline′s application in the synthesis of high-crystallinity polyaniline sheets, advancing the development of novel materials with potential uses in electronics and encryption technology (Li Z, 2024).

  • Borane-Trimethylamine Complex: A Versatile Reagent in Organic Synthesis: Aniline is crucial in the development of complex organic synthesis processes, demonstrating its utility in enhancing reagent capabilities within pharmaceutical manufacturing and chemical research (Perdicchia, 2024).


Analysis Note

Assay (according to weighing protocol): 725
Assay Cl (according to weighing protocol): 200
Assay (AOX): conforms

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service