Key Documents
EP6731010315
Eppendorf® CellXpert® C170i CO2 Incubator
new
water/humidity monitoring, inner door non-segmented, handle right side, AC/DC input 120 V, US 2-pin plug (US/JP/South America)
Synonym(s):
Heaters
About This Item
UNSPSC Code:
41104405
packaging
pack of 1 ea
manufacturer/tradename
Eppendorf® 6731010315
General description
CellXpert® C170i, non-segmented inner door, handle right side, water and humidity monitoring package, 100 – 127 V/50 – 60 Hz (US/JP/South America)
  • water and humidity monitoring package
  • handle right side
  • non-segmented inner door
  • 180° HTD (standard with C170i)
  • including water tray and four shelves (stainless steel)
  • 100 – 127 V, 50 – 60 Hz
Features and Benefits
  • water and humidity monitoring package
  • handle right side
  • non-segmented inner door
  • 180° HTD (standard with C170i)
  • including water tray and four shelves (stainless steel)
  • 100 – 127 V, 50 – 60 Hz
Legal Information
CellXpert is a registered trademark of Eppendorf AG
Eppendorf is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)