Skip to Content
Merck

AV31477

Anti-SLC30A9 (AB1) antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-Solute carrier family 30 (Zinc transporter), member 9

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.41
UNSPSC Code:
12352203
Conjugate:
unconjugated
Clone:
polyclonal
Application:
IHC, WB
Citations:
-
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

63 kDa

species reactivity

rat, human, horse, guinea pig, rabbit, bovine, dog

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable, western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SLC30A9(10463)

General description

SLC30A9 is a member of the solute carrier family that may function as a zinc transporter.
Rabbit Anti-SLC30A9 (AB1) antibody recognizes rat, human, canine, mouse, pig, bovine, and zebrafish SLC30A9.

The previously assigned protein identifier Q9Y6R2 has been merged into Q6PML9. Full details can be found on the UniProt database.

Immunogen

Synthetic peptide directed towards the N terminal region of human SLC30A9

Application

Rabbit Anti-SLC30A9 (AB1) antibody is suitable for use in western blot assays at a concentration of 0.5-2.0μg/ml. The antibody can also be used for IHC at 4-8μg/ml, using paraffin-embedded tissues.

Biochem/physiol Actions

The gene corresponding to embryonic lung protein [also known as Solute carrier family 30 (Zinc transporter), member 9, SLC30A9], is likely to be an evolutionarily conserved, housekeeping gene that plays a role intimately linked with cellular replication, DNA synthesis and/or transcriptional regulation.

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Other Notes

Synthetic peptide located within the following region: LKQEPLQVRVKAVLKKREYGSKYTQNNFITGVRAINEFCLKSSDLEQLR

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

10 - Combustible liquids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library