Select a Size
| Size/SKU | Availability | Price |
|---|
About This Item
biological source
rabbit
Quality Segment
conjugate
unconjugated
antibody form
affinity isolated antibody
antibody product type
primary antibodies
clone
polyclonal
purified by
affinity chromatography
species reactivity
human
technique(s)
immunocytochemistry: suitable, western blot: suitable
UniProt accession no.
shipped in
ambient
target post-translational modification
unmodified
Gene Information
human ... TACC3(10460)
General description
Immunogen
Application
Immunocytochemistry Analysis: A representative lot detected TACC3 spindle immunoreactivity in untransfected, but not TACC3 siRNA-transfected, SKOV-3 and OVCAR3 ovarian cancer cells (Rao, R., et al. (2014). Oncogene. 33(33):4242-4252).
Western Blotting Analysis: A representative lot detected much higher TACC3 expression in SKOV-3, OVCAR3 ovarian cancer cells than in non-cancerous hTERT-RPE, MCF10A, and 184B5 cells. Cellular TACC3 siRNA delivery abolished target band detection (Rao, R., et al. (2014). Oncogene. 33(33):4242-4252).
Epigenetics & Nuclear Function
Biochem/physiol Actions
Physical form
Preparation Note
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.
Analysis Note
Western Blotting Analysis: 1 µg/mL of this antibody detected TACC3 in 10 µg of HEK293 cell lysate.
Other Notes
Disclaimer
1 of 1
This Item | |||
|---|---|---|---|
| description from rabbit, purified by affinity chromatography | description affinity isolated antibody | description Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution | description - |
| conjugate unconjugated | conjugate unconjugated | conjugate unconjugated | conjugate unconjugated |
| antibody form affinity isolated antibody | antibody form affinity isolated antibody | antibody form affinity isolated antibody | antibody form - |
| Quality Level 100 | Quality Level 100 | Quality Level 100 | Quality Level 100 |
| biological source rabbit | biological source rabbit | biological source rabbit | biological source rabbit |
| shipped in ambient | shipped in wet ice | shipped in wet ice | shipped in wet ice |
| technique(s) immunocytochemistry: suitable, western blot: suitable | technique(s) ELISA: 1:1000, immunofluorescence: 1:100-1:500, immunohistochemistry: 1:50-1:100, western blot: 1:500-1:1000 | technique(s) immunofluorescence: 0.25-2 μg/mL, immunohistochemistry: 1:50-1:200, western blot: 0.04-0.4 μg/mL | technique(s) immunohistochemistry: 1:50-1:200, western blot: 1:500-1:2000 |
Still not finding the right product?
Try our Product Selector Tool to narrow your options
Storage Class
12 - Non Combustible Liquids
wgk
nwg
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.



