Merck
  • Detection of caffeine in tea, instant coffee, green tea beverage, and soft drink by direct analysis in real time (DART) source coupled to single-quadrupole mass spectrometry.
Detection of caffeine in tea, instant coffee, green tea beverage, and soft drink by direct analysis in real time (DART) source coupled to single-quadrupole mass spectrometry.
Journal of AOAC International (2013-06-19)
Lei Wang, Pengyue Zhao, Fengzu Zhang, Aijuan Bai, Canping Pan
ABSTRACT
Ambient ionization direct analysis in real time (DART) coupled to single-quadrupole MS (DART-MS) was evaluated for rapid detection of caffeine in commercial samples without chromatographic separation or sample preparation. Four commercial samples were examined: tea, instant coffee, green tea beverage, and soft drink. The response-related parameters were optimized for the DART temperature and MS fragmentor. Under optimal conditions, the molecular ion (M+H)+ was the major ion for identification of caffeine. The results showed that DART-MS is a promising tool for the quick analysis of important marker molecules in commercial samples. Furthermore, this system has demonstrated significant potential for high sample throughput and real-time analysis.
Product Description
Supelco
Melting point standard 235-237°C, analytical standard
Supelco
Caffeine solution, analytical standard, 1.0 mg/mL in methanol
Caffeine for system suitability, European Pharmacopoeia (EP) Reference Standard
Supelco
Caffeine solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
Caffeine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Caffeine Melting Point Standard, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Caffeine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Caffeine, anhydrous, 99%, FCC, FG
Supelco
Mettler-Toledo Calibration substance ME 18872, Caffeine, traceable to primary standards (LGC)
Sigma-Aldrich
Caffeine, BioXtra
Sigma-Aldrich
Caffeine, Sigma Reference Standard, vial of 250 mg
Sigma-Aldrich
Caffeine, powder, ReagentPlus®
Sigma-Aldrich
Caffeine, meets USP testing specifications, anhydrous
Sigma-Aldrich
Caffeine, anhydrous, tested according to Ph. Eur.
Caffeine, European Pharmacopoeia (EP) Reference Standard